5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals
5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 457410250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS628-46-6
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only