2,3-Diaminobenzoic acid, 95%, Thermo Scientific Chemicals
2,3-Diaminobenzoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Diaminobenzoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 457490010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS603-81-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to dark brown to black
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only