Thermo Scientific Chemicals

3-Acetylpyridine adenine dinucleotide, 90%, Thermo Scientific Chemicals

Catalog number: 457682500
250 mg, Each
Thermo Scientific Chemicals

3-Acetylpyridine adenine dinucleotide, 90%, Thermo Scientific Chemicals

Catalog number: 457682500
250 mg, Each
Quantity
Catalog number: 457682500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
86-08-8
IUPAC Name
3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium
Molecular Formula
C22H28N6O14P2
InChI Key
KPVQNXLUPNWQHM-RBEMOOQDSA-N
SMILES
CC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Appearance (Color)
White to off-white to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=89.0 %
Water
=<8 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text