Thermo Scientific Chemicals

Florisil™, 30-60 mesh, for column chromatography, Thermo Scientific Chemicals

Catalog number: 457735000
500 g, Each
Thermo Scientific Chemicals

Florisil™, 30-60 mesh, for column chromatography, Thermo Scientific Chemicals

Catalog number: 457735000
500 g, Each
Quantity
Catalog number: 457735000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1343-88-0
IUPAC Name
silicon(4+) magnesium(2+) trioxidandiide
Molecular Formula
MgO3Si
InChI Key
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)
100.39
Appearance
May contain black particles
Magnesium oxide (MgO)
15 to 16 %
Sodium sulfate (Na2SO4)
=<1.0
Appearance (Color)
White
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text