Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals
Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 457750250
Price (USD)/ Each
283.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
283.00
Add to cart
Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals
Catalog number457750250
Price (USD)/ Each
283.00
-
Add to cart
Chemical Identifiers
CAS89878-14-8
IUPAC Name3-(diethylboranyl)pyridine
Molecular FormulaC9H14BN
InChI KeyOJKBCQOJVMAHDX-UHFFFAOYSA-N
SMILESCCB(CC)C1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point164°C to 170°C
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only