Thermo Scientific Chemicals

1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals

Catalog number: 457770050
5 g, Each
Thermo Scientific Chemicals

1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific Chemicals

Catalog number: 457770050
5 g, Each
Quantity
Catalog number: 457770050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1143-38-0
IUPAC Name
1,8-dihydroxy-9,10-dihydroanthracen-9-one
Molecular Formula
C14H10O3
InChI Key
NUZWLKWWNNJHPT-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
Appearance (Color)
Yellow to red
Infrared spectrum
Conforms
Melting point
178°C to 182°C
HPLC
>=96.0 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text