Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate, 97%

Catalog number: 457941000
100 g, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate, 97%

Catalog number: 457941000
100 g, Each
Quantity
Catalog number: 457941000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15708-48-2
IUPAC Name
magnesium(2+) dipotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular Formula
C10H12K2MgN2O8
InChI Key
MUEOBEUHFKBRJH-UHFFFAOYSA-J
SMILES
[Mg++].[K+].[K+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Assay
96.0 to 104.0 %
pH
6.0 to 8.0 (5 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text