Coenzyme Q10, 98%
Coenzyme Q10, 98%
Coenzyme Q10, 98%
Thermo Scientific Chemicals

Coenzyme Q10, 98%

CAS: 303-98-0 | C59H90O4 | 863.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 457950010
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
-
Add to cart
Quantity:
1 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS303-98-0
IUPAC Name2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC59H90O4
InChI KeyACTIUHUUMQJHFO-UPTCCGCDSA-N
SMILESCOC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific absorbance A (1%/1cm)>=162 (275 nm, EtOH)
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472337Certificate of AnalysisJun 04, 2025457950010, 457950050, 457950250
A0444629Certificate of AnalysisFeb 03, 2025457950010, 457950050, 457950250
A0422991Certificate of AnalysisMay 02, 2023457950010, 457950050, 457950250
A0389266Certificate of AnalysisDec 12, 2022457950010, 457950050, 457950250
A0444629Certificate of AnalysisAug 04, 2022457950010, 457950050, 457950250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection