Thermo Scientific Chemicals

Coenzyme Q10, 98%

Catalog number: 457950250
25 g, Each
Thermo Scientific Chemicals

Coenzyme Q10, 98%

Catalog number: 457950250
25 g, Each
Quantity
Catalog number: 457950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
303-98-0
IUPAC Name
2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular Formula
C59H90O4
InChI Key
ACTIUHUUMQJHFO-UPTCCGCDSA-N
SMILES
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
Infrared spectrum
Conforms
HPLC
>=97.5 %
Specific absorbance A (1%/1cm)
>=162 (275 nm, EtOH)
Appearance (Color)
Yellow to orange
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text