Poly(vinyl alcohol), Thermo Scientific Chemicals
Poly(vinyl alcohol), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinyl alcohol), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
Catalog number 458230250
Price (USD)/ Each
33.65
Online exclusive
36.90 
Save 3.25 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
36.90 
Save 3.25 (9%)
Add to cart
Poly(vinyl alcohol), Thermo Scientific Chemicals
Catalog number458230250
Price (USD)/ Each
33.65
Online exclusive
36.90 
Save 3.25 (9%)
-
Add to cart
Chemical Identifiers
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
CAS9002-89-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Degree of hydrolysis99.3 to 100.0 mol %
pH5.5 to 7.5 (4 % in water)
Appearance (Color)White to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only