Thermo Scientific Chemicals

Poly(vinyl alcohol), Thermo Scientific Chemicals

Catalog number: 458245000
500 g, Each
Thermo Scientific Chemicals

Poly(vinyl alcohol), Thermo Scientific Chemicals

Catalog number: 458245000
500 g, Each
Quantity
Catalog number: 458245000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-89-5
Molecular Formula
(C2H4O)n
SMILES
OC(-*)C-*
MDL Number
MFCD00081922
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Assay
98 to 98.8 mol % (degree of hydrolysis)
Viscosity
3.5 to 4.5 cP (4 % in water)
Volatile matter
=<5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text