Thermo Scientific Chemicals

4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 458310010
1 g, Each
Thermo Scientific Chemicals

4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 458310010
1 g, Each
Quantity
Catalog number: 458310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
399-95-1
IUPAC Name
4-amino-3-fluorophenol
Molecular Formula
C6H6FNO
InChI Key
MNPLTKHJEAFOCA-UHFFFAOYSA-N
SMILES
NC1=CC=C(O)C=C1F
Appearance (Form)
Powder
GC
>=97.5 %
Appearance (Color)
Light yellow to brown
Melting point
137°C to 141°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text