4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals
4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 458310050
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
Add to cart
4-Amino-3-fluorophenol, 98%, Thermo Scientific Chemicals
Catalog number458310050
Price (USD)/ Each
207.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
244.00 
Save 36.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-amino-3-fluorophenol
Molecular FormulaC6H6FNO
InChI KeyMNPLTKHJEAFOCA-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C=C1F
Molecular Weight (g/mol)127.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=97.5 %
Appearance (Color)Light yellow to brown
Melting point137°C to 141°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only