Isothiazole, 97%, Thermo Scientific Chemicals
Isothiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isothiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458420010
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
Add to cart
Isothiazole, 97%, Thermo Scientific Chemicals
Catalog number458420010
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,2-thiazole
Molecular FormulaC3H3NS
InChI KeyZLTPDFXIESTBQG-UHFFFAOYSA-N
SMILESS1C=CC=N1
Molecular Weight (g/mol)85.12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only