Thermo Scientific Chemicals

4-Azaphthalide, 98%, Thermo Scientific Chemicals

Catalog number: 458450050
5 g, Each
Thermo Scientific Chemicals

4-Azaphthalide, 98%, Thermo Scientific Chemicals

Catalog number: 458450050
5 g, Each
Quantity
Catalog number: 458450050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5657-51-2
IUPAC Name
5H,7H-furo[3,4-b]pyridin-5-one
Molecular Formula
C7H5NO2
InChI Key
YSJHADWSLVFGGT-UHFFFAOYSA-N
SMILES
O=C1OCC2=C1C=CC=N2
Appearance (Color)
White to light brown
HPLC
>=97.5 %
Infrared spectrum
Conforms
Melting point
140°C to 146°C
Appearance (Form)
Powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text