Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals
Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
25 L
Catalog number 458480050
Price (USD)/ Each
-
Quantity:
5 L
Request bulk or custom format
Chemical Identifiers
CAS24800-44-0
IUPAC Name1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Molecular FormulaC9H20O4
InChI KeySNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILESCCC(O)OCC(C)OC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 % (sum of isomers)
Water=<0.05 %
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only