Thermo Scientific Chemicals

Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 458480250
25 L, Each
Thermo Scientific Chemicals

Tripropylene glycol, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 458480250
25 L, Each
Quantity
Catalog number: 458480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24800-44-0
IUPAC Name
1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Molecular Formula
C9H20O4
InChI Key
SNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILES
CCC(O)OCC(C)OC(O)CC
GC
>=98.5 % (sum of isomers)
Water
=<0.05 %
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text