Chemical Identifiers
CAS73590-58-6
IUPAC Name6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazole
Molecular FormulaC17H19N3O3S
InChI KeySUBDBMMJDZJVOS-UHFFFAOYNA-N
SMILESCOC1=CC=C2N=C(NC2=C1)S(=O)CC1=NC=C(C)C(OC)=C1C
View more