Thermo Scientific Chemicals

Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals

Catalog number: 458531000
100 mg, Each
Thermo Scientific Chemicals

Lithium tetrakis(pentafluorophenyl)borate ethyl etherate, Thermo Scientific Chemicals

Catalog number: 458531000
100 mg, Each
Quantity
Catalog number: 458531000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
371162-53-7
IUPAC Name
lithium(1+) ethoxyethane tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular Formula
C28H10BF20LiO
InChI Key
KPLZKJQZPFREPG-UHFFFAOYSA-N
SMILES
[Li+].CCOCC.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Appearance (Color)
White to light beige
Appearance (Form)
Powder or crystals
Melting point
117°C to 122°C
NMR (19F-NMR)
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text