Thermo Scientific Chemicals

Millon's reagent, for the detection of Tyrosine, Thermo Scientific Chemicals

Catalog number: 458545000
500 mL, Each
Thermo Scientific Chemicals

Millon's reagent, for the detection of Tyrosine, Thermo Scientific Chemicals

Catalog number: 458545000
500 mL, Each
Quantity
Catalog number: 458545000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10045-94-0
IUPAC Name
mercury(2+) dinitrate
Molecular Formula
HgN2O6
InChI Key
ORMNPSYMZOGSSV-UHFFFAOYSA-N
SMILES
[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Clear colorless to light yellow
Sensitivity
passes test
Density
1.2 to 1.5 kg/l
Composition
25 to 50 % (Nitric acid)
Composition
20 to 30 % (Mercury)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text