Thermo Scientific Chemicals

Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals

Catalog number: 458690025
2.5 L, Each
Thermo Scientific Chemicals

Ammonium hydroxide, 5% in water, Thermo Scientific Chemicals

Catalog number: 458690025
2.5 L, Each
Quantity
Catalog number: 458690025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1336-21-6
IUPAC Name
amine hydrate
Molecular Formula
H5NO
InChI Key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES
N.O
Color scale
=<10 APHA
Appearance (Color)
Clear solution
Strength
4.5 to 5.5 %
Residue after evaporation
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text