Iron(III) oxide, 95%, pure
Iron(III) oxide, 95%, pure
Iron(III) oxide, 95%, pure
Iron(III) oxide, 95%, pure
Thermo Scientific Chemicals

Iron(III) oxide, 95%, pure

CAS: 1309-37-1 | Fe2O3 | 159.69 g/mol
Have Questions?
Catalog NumberQuantity
4587000101 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458700010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to red-brown to rust brown
Titration Iodometric>=94 %
Loss on drying=<0.3 %
Appearance (Form)Powder
Ash=<0.3 % (1000°C, 0.5 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link