Thermo Scientific Chemicals

Amorolfine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 458750010
1 g, Each
Thermo Scientific Chemicals

Amorolfine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 458750010
1 g, Each
Quantity
Catalog number: 458750010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78613-38-4
IUPAC Name
hydrogen (2R,6S)-2,6-dimethyl-4-{2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl}morpholine chloride
Molecular Formula
C21H36ClNO
InChI Key
XZKWIPVTHGWDCF-KUZYQSSXSA-N
SMILES
[H+].[Cl-].CCC(C)(C)C1=CC=C(CC(C)CN2C[C@H](C)O[C@H](C)C2)C=C1
Appearance (Color)
White to light yellow to beige
Appearance (Form)
Crystalline powder or powder
Infrared spectrum
Conforms
Melting point
206°C to 211°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text