Chemical Identifiers
CAS78613-38-4
IUPAC Namehydrogen (2R,6S)-2,6-dimethyl-4-{2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl}morpholine chloride
Molecular FormulaC21H36ClNO
InChI KeyXZKWIPVTHGWDCF-KUZYQSSXSA-N
SMILES[H+].[Cl-].CCC(C)(C)C1=CC=C(CC(C)CN2C[C@H](C)O[C@H](C)C2)C=C1
View more