8-Bromoisoquinoline, 97%, Thermo Scientific Chemicals
8-Bromoisoquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromoisoquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 458760050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS63927-22-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light brown
Appearance (Form)Powder or crystalline powder
Melting point74°C to 82°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only