4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458820050
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
Add to cart
4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
Catalog number458820050
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Specifications
Chemical Name or Material4-Azabenzimidazole formate
Melting Point77.0°C to 78.0°C
Elemental Analysis48.87 to max. 52.95% min. (Carbon) 24.42 to max. 26.46% min. (Nitrogen)
Infrared SpectrumAuthentic
Molecular FormulaC6H5N3·CH2O2
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only