Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Abacavir sulfate, 99 %, Thermo Scientific Chemicals

Catalog number: 458860010
1 g, Each
Thermo Scientific Chemicals

Abacavir sulfate, 99 %, Thermo Scientific Chemicals

Catalog number: 458860010
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 458860010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
188062-50-2
IUPAC Name
bis([(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-en-1-yl]methanol); sulfuric acid
Molecular Formula
C28H38N12O6S
InChI Key
WMHSRBZIJNQHKT-FFKFEZPRSA-N
SMILES
OS(O)(=O)=O.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1
HPLC
>=98.5 %
Appearance (Form)
Crystalline powder
Water
=<1 %
Appearance (Color)
White to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text