1x1 image pixel for data collection
Abacavir sulfate, 99 %
Abacavir sulfate, 99 %
Abacavir sulfate, 99 %
Abacavir sulfate, 99 %
Thermo Scientific Chemicals

Abacavir sulfate, 99 %

CAS: 188062-50-2 | C14H18N61/2H2SO4 | 335.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458860050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS188062-50-2
IUPAC Namebis([(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-en-1-yl]methanol); sulfuric acid
Molecular FormulaC28H38N12O6S
InChI KeyWMHSRBZIJNQHKT-FFKFEZPRSA-N
SMILESOS(O)(=O)=O.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Water=<1 %
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link