Thermo Scientific Chemicals

Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 458870050
5 g, Each
Thermo Scientific Chemicals

Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 458870050
5 g, Each
Quantity
Catalog number: 458870050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
138-60-3
IUPAC Name
4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
Molecular Formula
C7H5NO5
InChI Key
XTLJJHGQACAZMS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=O)C=C(N1)C(O)=O
Water
=<10 %
Infrared spectrum
Conforms
Appearance (Color)
White to brown
HPLC
>=96.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text