Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals
Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 458870250
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
Add to cart
Chelidamic acid hydrate, 97%, Thermo Scientific Chemicals
Catalog number458870250
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
Molecular FormulaC7H5NO5
InChI KeyXTLJJHGQACAZMS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=O)C=C(N1)C(O)=O
Molecular Weight (g/mol)183.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
HPLC>=96.0 %
Water=<10 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only