Thermo Scientific Chemicals

cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 458950050
5 g, Each
Thermo Scientific Chemicals

cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 458950050
5 g, Each
Quantity
Catalog number: 458950050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
60662-54-6
GC
>=97.5 %
Appearance (Form)
Crystalline powder or crystals
Water
19 to 24 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text