Thermo Scientific Chemicals

Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals

Catalog number: 458970250
25 g, Each
Thermo Scientific Chemicals

Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals

Catalog number: 458970250
25 g, Each
Quantity
Catalog number: 458970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-03-8
IUPAC Name
propanoyl chloride
Molecular Formula
C3H5ClO
InChI Key
RZWZRACFZGVKFM-UHFFFAOYSA-N
SMILES
CCC(Cl)=O
Infrared spectrum
Conforms
Refractive index
1.4020 to 1.4050 (20°C, 589 nm)
Titration RCOCl
>=99.4 %
Appearance (Form)
Clear liquid
Color scale
=<20 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text