Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458975000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.90 
Save 9.25 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.90 
Save 9.25 (15%)
Add to cart
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Catalog number458975000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.90 
Save 9.25 (15%)
-
Add to cart
Chemical Identifiers
CAS79-03-8
IUPAC Namepropanoyl chloride
Molecular FormulaC3H5ClO
InChI KeyRZWZRACFZGVKFM-UHFFFAOYSA-N
SMILESCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4020 to 1.4050 (20°C, 589 nm)
Titration RCOCl>=99.4 %
Appearance (Form)Clear liquid
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only