Thermo Scientific Chemicals

Aluminum potassium sulfate, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 458990010
1 kg, Each
Thermo Scientific Chemicals

Aluminum potassium sulfate, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 458990010
1 kg, Each
Quantity
Catalog number: 458990010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-67-1
IUPAC Name
aluminium(3+) potassium disulfate
Molecular Formula
AlKO8S2
InChI Key
GRLPQNLYRHEGIJ-UHFFFAOYSA-J
SMILES
[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Heavy metals (as Pb)
=<0.003 %
Appearance (Color)
White
Appearance (Form)
Powder
Titration Complexometric
>=90 %
Loss on drying
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text