Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 459000010
Price (USD)
65.30
Each
In stock
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
65.30
Each
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Identification APasses test
Identification BPasses test
Identification CPasses test
Appearance (Color)White to light yellow
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473347Certificate of AnalysisJun 12, 2025459000050, 459000010
A0470572Certificate of AnalysisMay 17, 2025459000050, 459000010
A0468888Certificate of AnalysisJan 31, 2025459000050, 459000010
A0467230Certificate of AnalysisOct 31, 2024459000050, 459000010
A0464395Certificate of AnalysisJul 10, 2024459000050, 459000010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection