Thermo Scientific Chemicals

Iodomethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 459011000
100 g, Each
Thermo Scientific Chemicals

Iodomethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 459011000
100 g, Each
Quantity
Catalog number: 459011000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74-88-4
IUPAC Name
iodomethane
Molecular Formula
CH3I
InChI Key
INQOMBQAUSQDDS-UHFFFAOYSA-N
SMILES
CI
Appearance (Color)
Clear colorless to yellow to orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5290 to 1.5310 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text