Thermo Scientific Chemicals

2-Phenylacrylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 459050050
5 g, Each
Thermo Scientific Chemicals

2-Phenylacrylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 459050050
5 g, Each
Quantity
Catalog number: 459050050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
492-38-6
IUPAC Name
2-phenylprop-2-enoic acid
Molecular Formula
C9H8O2
InChI Key
ONPJWQSDZCGSQM-UHFFFAOYSA-N
SMILES
OC(=O)C(=C)C1=CC=CC=C1
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms
Melting point
104°C to 110°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text