7-Ethyl-10-hydroxycamptothecin, 98%, Thermo Scientific Chemicals
7-Ethyl-10-hydroxycamptothecin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Ethyl-10-hydroxycamptothecin, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459090010
Price (USD)/ Each
327.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
385.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
327.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
385.00 
Save 57.35 (15%)
Add to cart
7-Ethyl-10-hydroxycamptothecin, 98%, Thermo Scientific Chemicals
Catalog number459090010
Price (USD)/ Each
327.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
385.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
CAS86639-52-3
IUPAC Name(19S)-10,19-diethyl-7,19-dihydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular FormulaC22H20N2O5
InChI KeyFJHBVJOVLFPMQE-QFIPXVFZSA-N
SMILESCCC1=C2C=C(O)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Off-white to yellow
Appearance (Form)Powder
HPLC>=97.5 %
Water=<5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only