Chemical Identifiers
CAS139092-78-7
IUPAC Name4-(9H-carbazol-9-yl)-N,N-bis[4-(9H-carbazol-9-yl)phenyl]aniline
Molecular FormulaC54H36N4
InChI KeyAWXGSYPUMWKTBR-UHFFFAOYSA-N
SMILESC1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
View more