Thermo Scientific Chemicals

4-Iodo-1-trityl-1H-imidazole, 98%, Thermo Scientific Chemicals

Catalog number: 459200050
5 g, Each
Thermo Scientific Chemicals

4-Iodo-1-trityl-1H-imidazole, 98%, Thermo Scientific Chemicals

Catalog number: 459200050
5 g, Each
Quantity
Catalog number: 459200050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
96797-15-8
IUPAC Name
4-iodo-1-(triphenylmethyl)-1H-imidazole
Molecular Formula
C22H17IN2
InChI Key
DXJZJYPLPZEYBH-UHFFFAOYSA-N
SMILES
IC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
HPLC
>=97.5 %
Appearance (Color)
White to off-white
Appearance (Form)
Powder or solid
Melting point
222°C to 229°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text