Thermo Scientific Chemicals

L-Lysine methyl ester dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 459270050
5 g, Each
Thermo Scientific Chemicals

L-Lysine methyl ester dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 459270050
5 g, Each
Quantity
Catalog number: 459270050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26348-70-9
IUPAC Name
dihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
Molecular Formula
C7H18Cl2N2O2
InChI Key
SXZCBVCQHOJXDR-ILKKLZGPSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Specific optical rotation
+21° to +25° (20°C, 589 nm) (1% in MeOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text