Thermo Scientific Chemicals

(Pentafluoroethyl)trimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 459300010
1 g, Each
Thermo Scientific Chemicals

(Pentafluoroethyl)trimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 459300010
1 g, Each
Quantity
Catalog number: 459300010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124898-13-1
IUPAC Name
trimethyl(1,1,2,2,2-pentafluoroethyl)silane
Molecular Formula
C5H9F5Si
InChI Key
MTPVUVINMAGMJL-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C(F)(F)C(F)(F)F
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to almost colorless
Infrared spectrum
Conforms
Refractive index
1.3240 to 1.3260 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text