1x1 image pixel for data collection
L-Theanine, 98%
L-Theanine, 98%
L-Theanine, 98%
Thermo Scientific Chemicals

L-Theanine, 98%

CAS: 3081-61-6 | C7H14N2O3 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 459340010
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
In stock
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS3081-61-6
IUPAC Name2-amino-4-(ethylcarbamoyl)butanoic acid
Molecular FormulaC7H14N2O3
InChI KeyDATAGRPVKZEWHA-UHFFFAOYNA-N
SMILESCCNC(=O)CCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)white to off-white
Specific optical rotation+7.5° to +8.5° (20°C, 589 nm) (1 % in water)
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0460901Certificate of AnalysisMar 19, 2024459340050, 459340010
A0453144Certificate of AnalysisAug 25, 2023459340050, 459340010
A0448428Certificate of AnalysisDec 14, 2022459340050, 459340010
A0404900Certificate of AnalysisDec 13, 2022459340050, 459340010
A0413367Certificate of AnalysisDec 13, 2022459340050, 459340010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection