Chemical Identifiers
CAS24280-93-1
IUPAC Name(4E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydro-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
Molecular FormulaC17H20O6
InChI KeyHPNSFSBZBAHARI-RUDMXATFSA-N
SMILESCOC1=C(C)C2=C(C(=O)OC2)C(O)=C1C\C=C(/C)CCC(O)=O
View more