Thermo Scientific Chemicals

Mezlocillin sodium, 97%

Catalog number: 459560010
1 g, Each
Thermo Scientific Chemicals

Mezlocillin sodium, 97%

Catalog number: 459560010
1 g, Each
Quantity
Catalog number: 459560010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
42057-22-7
IUPAC Name
(2S,5R,6R)-6-[(2R)-2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium
Molecular Formula
C21H25N5NaO8S2
InChI Key
SGVORSZSYKDVFT-ZBJAFUORSA-N
SMILES
[Na].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
Appearance (Color)
White
Appearance (Form)
Powder and chunks
Infrared spectrum
Conforms
Water
=<6 %
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text