Chemical Identifiers
CAS42057-22-7
IUPAC Name(2S,5R,6R)-6-[(2R)-2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium
Molecular FormulaC21H25N5NaO8S2
InChI KeySGVORSZSYKDVFT-ZBJAFUORSA-N
SMILES[Na].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
View more