N,N-Diisopropylethylamine, 99.5+%, AcroSeal™, Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99.5+%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diisopropylethylamine, 99.5+%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number 459590010
Price (USD)/ Each
570.65
Online exclusive
634.00 
Save 63.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
570.65
Online exclusive
634.00 
Save 63.35 (10%)
Add to cart
N,N-Diisopropylethylamine, 99.5+%, AcroSeal™, Thermo Scientific Chemicals
Catalog number459590010
Price (USD)/ Each
570.65
Online exclusive
634.00 
Save 63.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7087-68-5
IUPAC Nameethylbis(propan-2-yl)amine
Molecular FormulaC8H19N
InChI KeyJGFZNNIVVJXRND-UHFFFAOYSA-N
SMILESCCN(C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
Refractive index1.4130 to 1.4150 (20°C, nm)
GC>=99.50 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only