Thermo Scientific Chemicals

Tin(II) chloride dihydrate, low in Mercury, suitable for AAS, Thermo Scientific Chemicals

Catalog number: 459602500
250 g, Each
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, low in Mercury, suitable for AAS, Thermo Scientific Chemicals

Catalog number: 459602500
250 g, Each
Quantity
Catalog number: 459602500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-69-1
IUPAC Name
λ²-tin(2+) dihydrate dichloride
Molecular Formula
Cl2H4O2Sn
InChI Key
FWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Sn++]
Copper (Cu)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text