Thermo Scientific Chemicals

Succinic acid disodium salt, Thermo Scientific Chemicals

Catalog number: 459635000
500 g, Each
Thermo Scientific Chemicals

Succinic acid disodium salt, Thermo Scientific Chemicals

Catalog number: 459635000
500 g, Each
Quantity
Catalog number: 459635000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-90-3
IUPAC Name
disodium butanedioate
Molecular Formula
C4H4Na2O4
InChI Key
ZDQYSKICYIVCPN-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<10 ppm
Titration with HClO4
98.5 to 102 %
UV
(0.1 M)
UV
at 280 nm A: =<0.1

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text