Thermo Scientific Chemicals

Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals

Catalog number: 459640010
1 g, Each
Thermo Scientific Chemicals

Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals

Catalog number: 459640010
1 g, Each
Quantity
Catalog number: 459640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
754-91-6
IUPAC Name
heptadecafluorooctane-1-sulfonamide
Molecular Formula
C8H2F17NO2S
InChI Key
RRRXPPIDPYTNJG-UHFFFAOYSA-N
SMILES
NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Infrared spectrum
Conforms
Appearance (Form)
Solid
GC
>=94 % (mix of isomers)
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text